Learn More
Magnesium sulfate heptahydrate, 99+%
Description
Magnesium sulfate, produced by appropriate processes, finds use in several pharmaceutical preparations of magnesium usually known as epsom salt. It is used as a micronutrient in agriculture, for brewing salt in the production of certain alcoholic products, as an electrolyte to prepare metal salts, as a coagulant and as an ingredient in bath salts for use in floatation therapy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Magnesium sulfate, produced by appropriate processes, finds use in several pharmaceutical preparations of magnesium usually known as epsom salt. It is used as a micronutrient in agriculture, for brewing salt in the production of certain alcoholic products, as an electrolyte to prepare metal salts, as a coagulant and as an ingredient in bath salts for use in floatation therapy.
Solubility
Soluble in in water and acid. Slightly soluble in alcohol and glycerol.
Notes
Incompatible with strong oxidizers.
Specifications
Specifications
| Quantity | 5000 g |
| Odor | Odorless |
| Density | 1.68 |
| Merck Index | 14,5691 |
| Solubility Information | Soluble in in water and acid. Slightly soluble in alcohol and glycerol. |
| Formula Weight | 246.48 (120.37 Anhydrous) |
| Percent Purity | ≥99% |
| Chemical Name or Material | Magnesium sulfate heptahydrate |
Safety and Handling
| EINECSNumber | 231-298-2 |
| RTECSNumber | OM4508000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only