missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate AR Crystal, Macron Fine Chemicals™
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 500 g |
| Packaging | POLYSTORMOR |
| Formula Weight | 246.47 |
| Percent Purity | 98.0 to 102.0% |
| Grade | ACS |
| Chemical Name or Material | Magnesium Sulfate, 7-Hydrate |