missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate Anhydrous, MilliporeSigma™

Catalog No. MMX00751
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX00751 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX00751 Supplier Technidata Supplier No. MX00751

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium(2+) sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 500 g
Linear Formula MgSO4
Formula Weight 12.37
Packaging Poly Bottle
Density 2.66g/cm3 (20°C); Bulk Density: 600 kg/m3
Chemical Name or Material Magnesium Sulfate, EMSURE™, For Analysis
Recommended Storage Store without restrictions