missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, 7-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 2.5 kg |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.67kg |
| Formula Weight | 246.47 |
| Percent Purity | 100.3% |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Chemical Name or Material | Magnesium Sulfate, 7-Hydrate |