missing translation for 'onlineSavingsMsg'
Learn More

Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3

Catalog No. AC196105000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196105000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196105000 Supplier Thermo Scientific Chemicals Supplier No. 196105000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7439-95-4 | Mg | 24.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy

Specifications

Chemical Name or Material Magnesium standard solution
Packaging Plastic bottle
Name Note 1 mg/ml Mg in 0.5N HNO3
CAS 7439-95-4, 7697-37-2, 7732-18-5
Density 1.74
Physical Form Liquid
Molecular Formula Mg
Concentration 997 to 1003 μg/mL of Mg (2+) in 0.5N HNO3 (20°C)
MDL Number MFCD00085308
Color Colorless
Fieser 16,198; 16,205; 16,356; 01,990; 04,422; 05,575; 06,507; 08,439; 10,345; 11,465; 14,281
Solubility Soluble in water
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES [Mg]
IUPAC Name magnesium
Molecular Weight (g/mol) 24.30
Quantity 500 mL
PubChem CID 5462224
ChEBI CHEBI:25107
Formula Weight 24.31
Grade AAS
Show More Show Less

RUO – Research Use Only