missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Magnesium peroxide complex

Catalog No. 433624250G
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
433624250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 433624250G Supplier Sigma Aldrich Supplier No. 433624250G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Magnesium peroxide complex

Specifications

CAS 1335-26-8
Linear Formula MgO2 -+ xMgO
Quantity 250 g
Synonym Magnesium dioxide