missing translation for 'onlineSavingsMsg'
Learn More

Magnesium lactate trihydrate, 99%

Catalog No. AA3936922
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3936922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3936922 Supplier Thermo Scientific Chemicals Supplier No. 03936922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18917-96-9 | C6H16MgO9 | 256.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18917-96-9
Molecular Formula C6H16MgO9
Molecular Weight (g/mol) 256.49
MDL Number MFCD00156534
Synonym magnesium lactate trihydrate, c6h10mgo6.3h2o, 2-hydroxypropanoyl oxy magnesio 2-hydroxypropanoate trihydrate
PubChem CID 91668363
IUPAC Name magnesium;2-hydroxypropanoate;trihydrate

Specifications

Melting Point 41°C (decomposition)
Color White
Quantity 100 g
Merck Index 14,5672
Solubility Information Soluble in water, Slightly soluble in alcohol
InChI Key SNLQXUYQWUDJLB-UHFFFAOYSA-L
SMILES CC(C(=O)[O-])O.CC(C(=O)[O-])O.O.O.O.[Mg+2]
Formula Weight 256.49 (202.45 Anhydrous)
Percent Purity 99%
Physical Form Powder or Lumps
Chemical Name or Material Magnesium lactate trihydrate
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 242-671-4
RTECSNumber OD5660000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only