missing translation for 'onlineSavingsMsg'
Learn More

Magnesium(II) Phthalocyanine, TCI America™

Catalog No. P10181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10181G Supplier TCI America Supplier No. P10181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1661-03-6
Molecular Formula C32H18MgN8O
Molecular Weight (g/mol) 554.856
MDL Number MFCD00041953
InChI Key HTNHYCHFBSVBID-UHFFFAOYSA-N
Synonym magnesium phthalocyanine
PubChem CID 91872988
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O.[Mg+2]

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 536.84
Physical Form Crystalline Powder
Chemical Name or Material Magnesium(II) Phthalocyanine
RTECSNumber OM4205000
TSCA Yes