missing translation for 'onlineSavingsMsg'
Learn More

Magnesium ICP-MS Standard, 1000 ppm Mg in 3% HNO3, Ricca Chemical

Catalog No. MSMG1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSMG1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSMG1KN100 Supplier Ricca Chemical Company Supplier No. MSMG1KN100

Chemical Identifiers

CAS 1309-48-4 , 7697-37-2 , 7732-18-5
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Mg
CAS Min % 93.0, 4.92, 0.15
CAS Max % 97.0, 5.12, 0.16
Equivalence 1 mL = 1 mg Mg
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Magnesium ICP-MS Standard
Show More Show Less