missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hydroxide, 95%, pure

Catalog No. AC315725000
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315725000 500 g Plastic bottle
AC315720250 25 g Plastic bottle
AC315720025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC315725000 Supplier Thermo Scientific Chemicals Supplier No. 315725000

CAS: 1309-42-8 | H2MgO2 | 58.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
SMILES [OH-].[OH-].[Mg++]

Specifications

Melting Point 350°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 94% min. (Complexometry)
Linear Formula Mg(OH)2
Merck Index 15, 5734
Solubility Information Solubility in water: insoluble
IUPAC Name magnesium(2+) dihydroxide
Formula Weight 58.33
Percent Purity 95%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Magnesium hydroxide, 95%
Show More Show Less
Hazard Category AIR SENSITIVE
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-170-3
RTECSNumber OM3570000
TSCA TSCA

RUO – Research Use Only