missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Reagent, For Total Cyanide, Certified, 51% (w/v), LabChem™

Catalog No. LC163901
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC163901 500 mL
LC163902 1 L
LC163904 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC163901 Supplier LabChem Supplier No. LC163901
Only null left

Chemical Identifiers

CAS 7791-18-6 , 7732-18-5
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Color Colorless
CAS Max % 54.52, 45.48
Linear Formula MgCl2·6H2O
Density 1.19g/mL
Solubility Information Soluble in water
Formula Weight 203.31
Percent Purity 51% (w/v)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.177g/mL ±0.005
Viscosity 2.7cSt
Decomposition Information Hydrogen chloride
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Magnesium Chloride Reagent, For Total Cyanide
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature