missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Hexahydrate, Fisher BioReagents
Description
Poly Bottle; 500g
Magnesium Chloride is suitable for use in the preparation of buffer solutions.
Specifications
Specifications
| Chemical Name or Material | Magnesium Chloride Hexahydrate |
| Melting Point | 117°C |
| CAS | 7791-18-6 |
| Color | White |
| pH | 5 to 6.5 |
| Physical Form | Powder/Solid |
| Quantity | 500 g |
| Packaging | Poly Bottle |
| Decomposition Information | >106°C |
| Assay Percent Range | ≥99.0 % |
| Show More |