missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate (Crystalline/USP/FCC/EP/BP), Fisher Chemical

Catalog No. M36212
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M36212 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M36212 Supplier Thermo Fisher Scientific Supplier No. M36212
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C
Color White
pH 5 to 6.5
Physical Form Powder/Solid
Quantity 2.5 kg
Assay Percent Range 99%
Linear Formula MgCl2·6H2O
Formula Weight 203.3
Percent Purity 98.0 to 101.0% (USP)
Grade BP/EP/FCC/USP
Identification Pass Test (BP), Pass Test (BP/EP/FCC/USP), Pass Test (EP), Pass Test (FCC), Pass Test (USP)
Packaging Poly Bottle
Appearance Clear and Colorless (EP)
Decomposition Information >106°C
Chemical Name or Material Magnesium Chloride Hexahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes and respiratory system. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1