missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Magnesium Chloride, Hexahydrate, Calbiochem™,

Catalog No. 442615500GM
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
442615500GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 442615500GM Supplier MilliporeSigma™ Supplier No. 442615500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS reagent grade magnesium chloride hexahydrate for research applications.

Hygroscopic

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Physical Form Crystalline Solid
Quantity 500 g
Solubility Information Easily soluble in cold water
Formula Weight 203.3
Grade ACS
Chemical Name or Material Magnesium Chloride, Hexahydrate
RTECSNumber OM2975000
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).