missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, ACS, 99.0-102.0%

Catalog No. AA36226A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36226A3 2 kg
AA3622636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36226A3 Supplier Thermo Scientific Chemicals Supplier No. 36226A3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol

Utilized in the polymerase chain reactions (PCR). Used for dust control, ice control, and road stabilization. Employed in a variety of applications such as fertilizer, mineral supplement for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Used in several medicines and topical or skin related applications. Acts as a coagulant in the preparation of tofu from soy milk. Acts as an anesthetic for cephalopods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C (decomposition)
Physical Form Crystalline
Quantity 2 kg
Odor Odorless
Sensitivity Hygroscopic
Assay Percent Range 99.0 to 102.0%
Density 1.570 g/mL
Merck Index 14,5662
Solubility Information Highly soluble in water and alcohol. Slightly soluble in acetone and pyridine.
Formula Weight 203.30 (95.22 Anhydrous)
Percent Purity 99.0 to 102.0%
Grade ACS
Chemical Name or Material Magnesium chloride hexahydrate, For analysis ACS
Show More Show Less
EINECSNumber 232-094-6
RTECSNumber OM2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only