missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride, 0.0100 M, Ricca Chemical

Catalog No. R44660001A
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R44660001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R44660001A Supplier Ricca Chemical Company Supplier No. R44660001A
Only null left

Chemical Identifiers

CAS 7791-18-6 , 7732-18-5
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Assay Percent Range 0.01000 ± 0.00005 M at 20°C
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solubility Infinitely soluble in water
Solvent or Matrix Water
Quantity 1 L
Chemical Name or Material Magnesium Chloride Aqueous Solutions, 0.01 M