missing translation for 'onlineSavingsMsg'
Learn More

Magnesium carbonate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013621
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013621 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013621 Supplier Merck Emd Millipore Supplier No. PHR13285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Magnesium carbonate is a naturally occurring magnesium salt. Magnesium carbonate can be prepared synthetically and is widely used in paints, printing ink, table salt and also as an antacid. It is also used in cosmetics as a colorling agent, perfume carrier and also as an anticaking agent.

Specifications

CAS 23389-33-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula MgCO3 · x H2O
MDL Number MFCD00151022
Synonym Magnesium carbonate; Magnesium carbonate hydrate
Type Certified Reference Material
Quantity 5 g
Formula Weight 84.31 (anhydrous basis)
Grade Certified Reference Material