missing translation for 'onlineSavingsMsg'
Learn More

Magnesium acetate, 1M aq. soln.

Catalog No. AAJ60041AE
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60041AE 100 mL
AAJ60041AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60041AE Supplier Thermo Scientific Chemicals Supplier No. J60041AE
Only null left

CAS: 142-72-3 | C4H6MgO4 | 142.393 g/mol

Magnesium acetate is used in molecular biology applications such as batch in vitro transcription of RNA and the crystallization of transcription factor:DNA complexes and proteins via the sitting-drop vapor-diffusion method. Also used as a buffer and to detect sodium and as a catalyst for sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium acetate is used in molecular biology applications such as batch in vitro transcription of RNA and the crystallization of transcription factor:DNA complexes and proteins via the sitting-drop vapor-diffusion method. Also used as a buffer and to detect sodium and as a catalyst for sulfuric acid.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 142-72-3
Molecular Formula C4H6MgO4
Molecular Weight (g/mol) 142.393
MDL Number MFCD00003539
InChI Key UEGPKNKPLBYCNK-UHFFFAOYSA-L
Synonym magnesium acetate, magnesium diacetate, cromosan, acetic acid, magnesium salt, mg acetate, magnesium di acetate, nu-mag, unii-0e95jzy48k, ccris 7883, acetic acid, magnesium salt 2:1
PubChem CID 8896
ChEBI CHEBI:62964
IUPAC Name magnesium;diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Mg+2]

Specifications

Physical Form Liquid
Quantity 100 mL
Linear Formula Mg(CH3COO)2
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 142.39
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Chemical Name or Material Magnesium acetate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only