Learn More
Magnesium AA Standard, Certified, 1000ppm Mg ±10ppm (1mL = 1mg Mg), LabChem™
Supplier: LabChem LC164801
Specifications
| Magnesium AA Standard | |
| Poly Bottle | |
| 96.49,2.46,1.05 | |
| 1g/mL | |
| H12MgN2O12 | |
| Mg(NO3)2·6(H2O) | |
| UN3264 | |
| Soluble in water | |
| O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 256.40 | |
| 202877 | |
| Certified |
| 1000ppm (1mL = 1mg Mg) | |
| 13446-18-9 , 7697-37-2 , 13446-18-9 | |
| Passes Test | |
| Liquid | |
| 1000 ppm Mg ±10 ppm (1mL = 1mg Mg) | |
| MFCD00149779 | |
| Colorless | |
| MFUVDXOKPBAHMC-UHFFFAOYSA-N | |
| magnesium(2+) hexahydrate dinitrate | |
| 500 mL | |
| 256.41 |
Chemical Identifiers
| 13446-18-9 | |
| 256.40 | |
| MFUVDXOKPBAHMC-UHFFFAOYSA-N | |
| magnesium(2+) hexahydrate dinitrate |
| H12MgN2O12 | |
| MFCD00149779 | |
| 202877 | |
| O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature