missing translation for 'onlineSavingsMsg'
Learn More

Magnesium AA Standard, Certified, 1000ppm Mg ±10ppm (1mL = 1mg Mg), LabChem™

Catalog No. p-5388663
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC164807 125 mL Poly Bottle
LC164801 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC164807 Supplier LabChem Supplier No. LC164807
Only null left

Chemical Identifiers

CAS 13446-18-9 , 7697-37-2 , 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
PubChem CID 202877
IUPAC Name magnesium(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly Bottle
CAS Max % 96.49, 2.46, 1.05
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm Mg ±10 ppm (1mL = 1mg Mg)
Linear Formula Mg(NO3)2·6(H2O)
UN Number UN3264
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 256.41
Grade Certified
Chemical Name or Material Magnesium AA Standard, 1000ppm (1mL = 1mg Mg)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature