missing translation for 'onlineSavingsMsg'
Learn More

Magnesium AA Standard, 1000 ppm Mg in 3% HCl, Ricca Chemical

$280.34 - $280.34

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AMG1KH-500
View Documents
Ricca Chemical Company
AMG1KH500
500 mL Natural Poly Bottle
Each for $280.34
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1309-48-4
40.30
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
MgO
MFCD00011109
14792
O=[Mg]

Specifications

Natural Poly Bottle
94.0,4.2,0.16
97.0,4.37,0.16
Liquid
MFCD00011109
100°C
O=[Mg]
40.30
14792
1309-48-4,7647-01-0,7732-18-5
1,000ppm Mg
1 mL = 1 mg Mg
MgO
Colorless
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
500 mL
Magnesium AA Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'