missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium AA Standard, 1000 ppm Mg in 3% HCl, Ricca Chemical
Supplier: Ricca Chemical Company AMG1KH500
Specifications
| Magnesium AA Standard | |
| 1309-48-4,7647-01-0,7732-18-5 | |
| 1,000ppm Mg | |
| 1 mL = 1 mg Mg | |
| MgO | |
| Colorless | |
| CPLXHLVBOLITMK-UHFFFAOYSA-N | |
| oxomagnesium | |
| 500 mL |
| Natural Poly Bottle | |
| 94.0,4.2,0.16 | |
| 97.0,4.37,0.16 | |
| Liquid | |
| MFCD00011109 | |
| 100°C | |
| O=[Mg] | |
| 40.30 | |
| 14792 |
Chemical Identifiers
| 1309-48-4 | |
| 40.30 | |
| CPLXHLVBOLITMK-UHFFFAOYSA-N | |
| oxomagnesium |
| MgO | |
| MFCD00011109 | |
| 14792 | |
| O=[Mg] |