missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagnaBind™ Protein G Beads

Catalog No. PI21349
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21349 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21349 Supplier Thermo Scientific™ Supplier No. 21349

Enable benchtop magnetic affinity purification with these ligand-coated (streptavidin, Protein A and G, GAM IgG, GAR IgG) magnetic particles and magnet separators.

Thermo Scientific™ MagnaBind™ Beads provide a convenient method for magnetic separation of antibodies, antigens, lectins, enzymes, nucleic acids and cells using affinity binding. To remove the MagnaBind Beads from the suspension, an external magnetic field is used.

Features of MagnaBind Protein G Beads:

  • Composition: Silanized iron oxide
  • Magnetization: 25-35EMU/g
  • Type of Magnetization: Superparamagnetic (no magnetic memory)
  • Surface Area: >100m2/g
  • Bead Size: 1-4μm diameter
  • Settling Rate: 4% in 30 minutes
  • Effective Density: 2.5g/mL
  • Number of Beads: 1 x 108 beads/mg
  • pH Stability: Aqueous solution, above pH 4.0
  • Concentration: 5 mg/mL

MagnaBind Protein G Beads are typically used to isolate antibodies from serum and cell culture supernatant and to separate cells of interest from a cell mixture. For antibody purification, the beads are incubated with the antibody solution and then magnetically separated from the supernatant. The antigens and antibodies are then dissociated from the beads using an elution buffer. For cell separation, the monoclonal or polyclonal antibody to the cell surface antigen is incubated with Protein G magnetic beads, magnetically separated, and then incubated with the cell suspension.

Applications:

  • Cell sorting using either positive or negative selection
  • Protein purification or immunoassays using either direct or indirect methods

Specifications

Capacity (Metric) 5 mL
Concentration Slurry: 5 mg/mL
Description MagnaBind Protein G Beads
Content And Storage Upon receipt store products at 4°C. Do not freeze products. Products are shipped at ambient temperature.
Particle Size 1 to 4 μm
Form Liquid
Product Line MagnaBind
Quantity 5 mL
Target Antibody
Type Magnetic Affinity Bead
Ligand Type Protein G
Sufficient For 0.2 mg of Mouse IgG per mL of Beads
Show More Show Less
What are the differences between Protein A, Protein G, Protein A/G, and Protein L Ig-binding proteins?

Protein A, Protein G, and Protein A/G bind almost exclusively to the IgG class of antibodies, but their binding properties differ among species and subclasses of IgG. Protein L binds in the variable fragment of some kappa light chains and can react with any immunoglobulin, not just IgG, as long as the correct kappa light chains are present. Protein L does not bind lambda light chains and certain kappa chains of different species.

-Protein A is generally preferred for rabbit, pig, dog, and cat IgG.
-Protein G has better binding capacity for a broader range of mouse and human IgG subclasses (e.g., IgG1 vs. IgG2)
-Protein A/G is a recombinant fusion protein that includes the IgG-binding domains of both Protein A and Protein G. Therefore, Protein A/G is ideal for binding the broadest range of IgG subclasses from rabbit, mouse, human, and other mammalian samples.
-Protein L binds to certain immunoglobulin kappa light chains. Because kappa light chains occur in members of all classes of immunoglobulin (i.e., IgG, IgM, IgA, IgE and IgD), Protein L can purify these different classes of antibody. However, only those antibodies within each class that possess the appropriate kappa light chains will bind. Generally, empirical testing is required to determine if Protein L is effective for purifying a particular antibody. It binds only Vk1 in mouse and VkI, VkIII and VkIV in human.
Read more about the general characteristics of Ig-binding proteins (https://www.thermofisher.com/us/en/home/life-science/antibodies/antibody-purification-kits-reagents.html) and (https://assets.thermofisher.com/TFS-Assets/LSG/Application-Notes/TR0034-Ab-binding-proteins.pdf).

For Research Use Only. Not for use in diagnostic procedures.