missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Magna ChIP™ Protein AG Magnetic Beads

Catalog No. 16663
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16663 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16663 Supplier MilliporeSigma™ Supplier No. 16663

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Capacity (Metric) 1 mL
Content And Storage 2°C to 8°C. Do not freeze.
Particle Size ∼ 3 μm
Quantity 1 mL
For Use With (Application) Chromatin immunoprecipitations (ChIP) and RNA immunoprecipitations (RIP) assays
Type Magnetic Affinity Bead
Sufficient For 50 Reactions