missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Microbiome Bead Plate

Catalog No. A42331
Encompass
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42331 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42331 Supplier Applied Biosystems™ Supplier No. A42331
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This bead plate is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.

This bead plate is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. No. A42357). It is made available separately for applications that require additional plates.

Specifications

Content And Storage Store at room temperature.

Sample Type Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
Product Line MagMAX
Quantity 1 plate
Shipping Condition Room Temperature
Target Viral DNA/RNA, Pathogen DNA/RNA, Total Nucleic Acids

For Research Use Only. Not for use in diagnostic procedures.