missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagicMark™ XP Western Protein Standard

Numéro de catalogue. p-4924116
Encompass
Change view
Click to view available options
Quantity:
50 μL
250 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
LC5602 250 μL
LC5603 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. LC5602 Fournisseur Invitrogen™ Code fournisseur LC5602
Il en reste null

Comprend

250 μL of ready-to-use standard mixture is provided in a plastic vial; Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue

Specifically designed for easy and convenient protein molecular weight estimation directly on Western blots

  • MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site
  • The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot
  • The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent)
  • The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use
  • Store at -20°C

Applications

  • Western blotting: detection of the nine unstained bands via the detection method used for the target protein. Compatible with chemiluminescent substrates and fluorescent secondary antibodies (not recommended for antibodies labeled with fluors in the 500–550 nm channel)
  • Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method

Order Info

Shipping Condition: Dry Ice

Spécifications

Content And Storage 250 μL of ready-to-use standard mixture is provided in a plastic vial. Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue.

Store at -20°C.
Detection Method User defined detection system
Number of Markers 9
Ready to Load Yes
Size Range 20 to 220 kDa
Gel Compatibility Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 220, 120, 100, 80, 60, 50, 40, 30, 20 kDa
Quantity 250 μL
Shipping Condition Dry Ice
Product Line MagicMark
Product Type Protein Ladder
Stain Type Unstained
System Type Western Blotting, SDS-PAGE
Afficher plus de résultats Afficher moins de résultats

For Research Use Only. Not for use in diagnostic procedures.