missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals MagBeads FastRNA Kit for Blood

Catalog No. MP116586050
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116586050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116586050 Supplier MP Biomedicals Supplier No. 116586050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Efficiently purify high-quality RNA from whole blood with a simplified, lysis-free workflow. Removes DNA contamination for pure RNA, and supports scalable, automated processing for high-throughput applications.

  • MagBeads FastRNA™ Kit for Blood is a scalable and automatable (automatable versions available cat. no.117039100 and cat. no.117039200) magnetic bead-based RNA purification technology that is designed for total isolation of RNA from whole blood preserved in different anticoagulants (EDTA, Heparin and sodium citrate). RNA degradation prevention is the backbone of RNA purification. The use of our specially formulated Buffer MBL containing huge amounts of chaotropic ions omits the need of excessive homogenization and enables highly efficient lysis with the immediate inactivation of RNases, which are typically present in most biological materials.
  • The MagBeads FastRNA™ Kit for Blood simplifies the RNA purification process by eliminating the need for complicated erythrocyte lysis and isolation of leukocyte pellet, making it hassle-free and time efficient. Contaminated DNA is removed by a DNase I treatment step. Simplified dual washing steps largely remove salts, metabolites, and macromolecular cellular components. Pure RNA extracts of high yield and purity, compatible with downstream applications such as RT-qPCR amplification, are then eluted under low ionic strength conditions with RNase-free water.

Specifications

Form Liquid
Quantity 50 Preps
Type Magnetic Affinity Bead
Sufficient For Blood