missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals MagBeads FastDNA Kit for Soil

Catalog No. MP119607096
Change view
Click to view available options
No. of Reactions:
96 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP119607096 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP119607096 Supplier MP Biomedicals Supplier No. 119607096

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Delivers high-quality genomic DNA from soil samples in under 60 minutes with efficient processing workflow.

  • The MagBeads FastDNA™ Kit for Soil allows quick and efficient isolation of high-quality genomic DNA from soil in less than 60 mins
  • Samples are placed into Lysing Matrix E tube and used with FastPrep™ Instruments from MP Biomedicals to effectively lyse host cells as well as bacteria, fungi, viruses, protists, and other cells present in soil samples within 40 seconds
  • The kit is also compatible with most of the automated nucleic acid extraction instruments on the market or it can be operated manually
  • It consists of specially formulated reagents to eliminate humic acid, polysaccharides, phenolic compounds, and enzyme inhibitors from soil and thus allowing extraction of highly pure genomic DNA ready for PCR, restriction digestion, electrophoresis, and other desired applications

Specifications

Format Kit
Isolation Technology Magnetic Bead
Scale Mini
No. of Reactions 96 Preps
Product Type MagBeads FastDNA Kit
Voltage 0/1 V