missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals MagBeads FastDNA™ Kit for Microbiome
Description
- The MagBeads FastDNA Kit for Microbiome is an advanced genomic DNA (gDNA) extraction solution based on the next-generation silicon-based magnetic bead technology.
- This kit is designed to deliver high-fidelity gDNA extraction from a diverse array of biological specimens, encompassing bacterial communities, a spectrum of bodily fluids, bronchoalveolar lavage fluids, and environmental samples such as soil
- The kit integrates an effective lysis mechanism, ensuring thorough cell lysis and subsequent DNA purification across a broad spectrum of microorganisms, including bacteria, archaea, and fungi.
- The resulting gDNA is of high purity and suitable for numerous downstream molecular applications, such as Polymerase Chain Reaction (PCR), Quantitative Real-Time PCR (qRT-PCR), and metagenomic library construction, facilitating comprehensive genomic analyses.
Specifications
Specifications
| Form | Liquid |
| Quantity | 50 Preps |
| Type | Magnetic Affinity Bead |
| Sufficient For | Microbiome |