missing translation for 'onlineSavingsMsg'
Learn More

MafA Rabbit anti-Mouse, Polyclonal, Bethyl Laboratories

Numéro de catalogue. 501558041
Encompass
Modifier l'affichage
Click to view available options
Quantité:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
501558041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 501558041 Fournisseur Bethyl Laboratories Inc Code fournisseur A300611A

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Human

MAFA is a transcription factor that binds RIPE3b, a conserved enhancer element that regulates pancreatic beta cell-specific expression of the insulin gene. MAFA cooperates synergistically with NEUROD1 and PDX1. Phosphorylation by GSK3 increases its transcriptional activity and is required for its ocogenic activity. Serves as an ocogene or tumor supressor depending on the cell context.

Spécifications

Antigène MafA
Applications Immunoelectrophoresis, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugué Unconjugated
Formule phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gène Mafa
Numéro d’acquisition du gène Q8CF90
Alias du gène RIPE3b1, hMafA, pancreatic beta-cell-specific transcriptional activator, transcription factor MafA, transcription factor RIPE3b1
Symboles de gène Mafa
Espèces hôtes Rabbit
Immunogène Between 300 and C-term
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 378435
Espèces cibles Mouse
Contenu et stockage 4° C
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats