Learn More
Mopec Maestro Recirculating to In-House Facility Vented Conversion Kit
Designed to convert recirculating grossing stations into in-house ventilation
Supplier: Mopec AX110060
Description
- Designed to convert recirculating grossing stations into in-house ventilation
- Bolt-on panel with 8 in duct stubs which allows the grossing station to be connected to the facility ventilation system
- Includes panel, duct tubing, and clamps
- 304 stainless steel sheetmetal and hardware
- Fits the Mopec Maestro and Maestro Encore grossing stations of the accompanying sized unit
Identifiants chimiques
| Grossing Station | |
| Stainless Steel (304) |
Spécifications
| Grossing Station | |
| Stainless Steel (304) | |
| 60 in. |