missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Agar (Dehydrated)

Numéro de catalogue. p-10710000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
OXCM0007B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. OXCM0007B Fournisseur Thermo Scientific™ Code fournisseur CM0007B
Il en reste null

Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with this chromogenic medium.

Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with Thermo Scientific™ Oxoid™ MacConkey Agar (Dehydrated). This differential medium gives a number of diagnostic indications such as bile tolerance, colony morphology and chromogenesis. MacConkey Agar corresponds to the medium recommended by the World Health Organizaton1, the Department of Health2 for the bacterial examination of water.

Use MacConkey Agar for the cultivation of pathogens which may be present in a variety of specimens.

  • Easy-to-read: easy to differentiate Escherichia coli (red non-mucoid colonies) from Staphylococcus spp. (pale pink opaque colonies) and Pseudomonas aeruginosa (green-brown) colonies.
  • Recommended by World Health Organizaton1, the Department of Health2.

Due to its ability to support the growth of pathogenic Gram-positive cocci (e.g. staphylococci and enterococci) as well as Enterobacteriaceae, MacConkey Agar is particularly recommended for the cultivation of pathogens which may be present in a variety of specimens such as urine, feces and wound swabs. While it is selective it does not suppress a mixed bacterial flora to the same extent as other inhibitory media (including other MacConkey agars). MacConkey Agar should be used in parallel with other selective indicator media such as Desoxycholate Citrate Agar, Bismuth Sulphite Agar, Brilliant Green Agar and Brilliant Green Bile (2%) Broth, and a non-selective medium such as Blood Agar.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Spécifications

Description MacConkey Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 9.6L medium