Learn More
Mettler Toledo™ M1 OIML Class Standard Weight (M1 PL E)
Single M1 OIML weight constructed of 304 stainless steel and housed in a plastic box.
$27.69 - $1758.67
Specifications
Product Type Test Weight
Material Stainless Steel
Case Material Plastic
Density 7950 (± 140) kg/m3
Includes Without certificate
View More Specs
Products 23
Catalog Number Mfr. No. Weight (Metric) Shape Price Quantity  
Catalog Number Mfr. No. Weight (Metric) Shape Price Quantity  
01912649
View Documents
Mettler Toledo™
30402184
1 mg Sheet
Each for $27.69
Only null left
01912650
View Documents
Mettler Toledo™
30402510
2 mg Sheet
Each for $27.90
Only null left
01912651
View Documents
Mettler Toledo™
30402355
5 mg Sheet
Each for $27.69
Only null left
01912652
View Documents
Mettler Toledo™
30402585
10 mg Sheet
Each for $27.90
Only null left
01912653
View Documents
Mettler Toledo™
30402586
20 mg Sheet
Each for $27.90
Only null left
01912654
View Documents
Mettler Toledo™
30402587
50 mg Sheet
Each for $27.69
Only null left
01912655
View Documents
Mettler Toledo™
30402638
100 mg Sheet
Each for $27.90
Only null left
01912656
View Documents
Mettler Toledo™
30402639
200 mg Sheet
Each for $27.90
Only null left
01912657
View Documents
Mettler Toledo™
30402640
500 mg Sheet
Each for $49.12
Only null left
01912658
View Documents
Mettler Toledo™
30402641
1 g Knob with Adjusting Cavity
Each for $33.45
Only null left
01912659
View Documents
Mettler Toledo™
30402642
2 g Knob with Adjusting Cavity
Each for $33.45
Only null left
01912660
View Documents
Mettler Toledo™
30402643
5 g Knob with Adjusting Cavity
Each for $41.54
Only null left
01912661
View Documents
Mettler Toledo™
30402644
10 g Knob with Adjusting Cavity
Each for $54.99
Only null left
01912662
View Documents
Mettler Toledo™
30402645
20 g Knob with Adjusting Cavity
Each for $60.54
Only null left
01912663
View Documents
Mettler Toledo™
30402646
50 g Knob with Adjusting Cavity
Each for $91.87
Only null left
01912664
View Documents
Mettler Toledo™
30402647
100 g Knob with Adjusting Cavity
Each for $108.15
Only null left
01912665
View Documents
Mettler Toledo™
30402576
200 g Knob with Adjusting Cavity
Each for $120.78
Only null left
01912666
View Documents
Mettler Toledo™
30402699
500 g Knob with Adjusting Cavity
Each for $125.84
Only null left
01912667
View Documents
Mettler Toledo™
30402702
1 kg Knob with Adjusting Cavity
Each for $195.07
Only null left
01912668
View Documents
Mettler Toledo™
30402665
2 kg Knob with Adjusting Cavity
Each for $251.67
Only null left
01912669
View Documents
Mettler Toledo™
30402708
5 kg Knob with Adjusting Cavity
Each for $591.28
Only null left
01912670
missing translation for 'viewDocuments'
Mettler Toledo™
30402711
10 kg Knob with Adjusting Cavity
Each for $1,258.36
Only null left
01912671
missing translation for 'viewDocuments'
Mettler Toledo™
30402780
20 kg Knob with Adjusting Cavity
Each for $1,758.67
Only null left
Description
Description
  • Without calibration certificate
  • For Standard Enviroments: Affordable quality weights for regular routine testing without the need for traceability
  • Weights with knob design have an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment
Specifications
Specifications
Test Weight
Plastic
Without certificate
Stainless Steel
7950 (± 140) kg/m3