missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Xylène 99,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Quantity | 25 mL |
| Physical Form | Liquide clair à 20 °C |
| UN Number | 1307 |
| Boiling Point | 139°C |
| Formula Weight | 106.17 |
| Percent Purity | ≥99.0% (GC) |
| Chemical Name or Material | m-Xylene |
Sécurité et manipulation
| EINECSNumber | (3)-0003&(3)-0060 |
| RTECSNumber | ZE2275000 |
| TSCA | Yes |