missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. T03175G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T03175G Supplier TCI America Supplier No. T03175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 637-04-7
Molecular Formula C7H11ClN2
Quantity 5 g
MDL Number MFCD00012932
Synonym 3-methylphenylhydrazine hydrochloride, m-tolylhydrazine hydrochloride, m-tolylhydrazinium 1+ chloride, 3-methylphenyl hydrazine hydrochloride, 3-methylphenylhydrazine hcl, m-tolyhydrazine hydrochloride, 1-3-methylphenyl hydrazine hydrochloride, 3-tolylhydrazine hydrochloride, 3-methyl phenylhydrazine hydrochloride, hydrazine, 3-methylphenyl-, monohydrochloride
InChI Key SHTBTGXNVWYODU-UHFFFAOYSA-N
SMILES [H+].[Cl-].CC1=CC=CC(NN)=C1
IUPAC Name hydrogen (3-methylphenyl)hydrazine chloride
Molecular Weight (g/mol) 158.63
PubChem CID 71562
Formula Weight 158.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material m-Tolylhydrazine Hydrochloride
Show More Show Less
EINECSNumber (3)-0468
TSCA No