missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ M-PER™ Mammalian Protein Extraction Reagent

Numéro de catalogue. p-4531885
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PI78501 250 mL
PI78503 25 mL
PI78505 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. PI78501 Fournisseur Thermo Scientific™ Code fournisseur 78501
Il en reste null

Isolate total protein from cultured mammalian cells with this highly efficient lysis and extraction reagent.

Thermo Scientific M-PER Mammalian Protein Extraction Reagent is designed to provide highly efficient total soluble protein extraction from cultured mammalian cells.

Features of M-PER Mammalian Protein Extraction Reagent:

  • Gentle—mild detergent lysis, yielding extracts that are immediately compatible with coomassie (Bradford) and BCA protein assays or SDS-PAGE
  • Compatible—extracts soluble proteins in nondenatured state, enabling direct use in immunoprecipitation and other affinity purification procedures
  • Amine-free and dialyzable—formulation ensures compatibility with subsequent assay systems
  • Convenient—lyse adherent cells directly in plate or after scraping and washing in suspension
  • Non-denaturing—maintain activity of luciferase, beta-galactosidase, CAT and other reporter genes as well or better than other suppliers' products and freeze/thaw methods

Product Details

The complete cell lysis reagent contains a mild, nondenaturing detergent that dissolves cell membranes to extract and solubilize total protein from most cellular compartments. Extraction is accomplished in only 5 minutes and requires little or no additional mechanical disruption. M-PER reagent is formulated for minimal interference with downstream biological applications. The reagent has been validated for use with several cell lines, including primary, suspension and adherent cell types; the resulting cell lysates are compatible with many downstream assays including immunoassays, enzyme assays and a variety of common reporter assays.

Cellular protein extraction—cell lysis to release the proteins of interest—is a key first step in many proteomics analysis procedures. M-PER reagent is designed to efficiently extract soluble protein from a variety of cell types, including primary cells and cells grown in suspension or adherent culture conditions. Performance of M-PER reagent was evaluated for yields of both total protein and specific target proteins from the various cellular compartments. Additionally, because M-PER reagent is formulated with dialyzable components that are devoid of primary amines, it is more compatible with certain downstream applications, such as luciferase, beta-galactosidase, and CAT assays, than other protein extraction methods.

Related Products

  • T-PER™ Tissue Protein Extraction Reagent (Cat. No. 78510)

Spécifications

Quantity 250 mL
Format Liquid
Product Type Protein Extraction Reagent
Volume (Metric) 250 mL
Content And Storage Upon receipt store product at room temperature.
Product Line M-PER

For Research Use Only. Not for use in diagnostic procedures.