missing translation for 'onlineSavingsMsg'
Learn More

m-Cymene 99.0+%, TCI America™

Catalog No. C07981ML
Change view
Click to view available options
Quantity:
0.1 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07981ML 1 mL
C079801ML 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07981ML Supplier TCI America Supplier No. C07981ML
Only null left

Chemical Identifiers

CAS 535-77-3
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00008891
InChI Key XCYJPXQACVEIOS-UHFFFAOYSA-N
Synonym m-cymene, 1-isopropyl-3-methylbenzene, 3-isopropyltoluene, m-isopropyltoluene, m-cymol, m-methylisopropylbenzene, beta-cymene, benzene, 1-methyl-3-1-methylethyl, 1-methyl-3-isopropylbenzene, 1-methyl-3-1-methylethyl benzene
PubChem CID 10812
IUPAC Name 1-methyl-3-(propan-2-yl)benzene
SMILES CC(C)C1=CC=CC(C)=C1

Specifications

Color Colorless
Boiling Point 175°C
Quantity 1 mL
UN Number 2046
Formula Weight 134.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material m-Cymene
EINECSNumber (3)-0012&(3)-0015
RTECSNumber DA6127310
TSCA Yes