missing translation for 'onlineSavingsMsg'
Learn More

m-Carborane, 98%, Thermo Scientific Chemicals

Numéro de catalogue. p-7059786
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA4560609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AA4560609 Fournisseur Thermo Scientific Chemicals Code fournisseur 04560609
Il en reste null

m-Carborane is used as a pharmaceutical, chemical and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 16986-24-6
Molecular Formula C2H12B10
Molecular Weight (g/mol) 144.25
MDL Number MFCD00151574
InChI Key JTWJUVSLJRLZFF-UHFFFAOYSA-N
Synonym m-carborane, 1,7-dicarbadodecaborane 12, 2,3,4,5,6,7,8,9,11,12-decaborabicyclo 8.1.1 dodecane
PubChem CID 16212388
IUPAC Name 2$l^{2},3$l^{2},4$l^{2},5$l^{2},6$l^{2},7$l^{2},8$l^{2},9$l^{2},11$l^{2},12$l^{2}-decaborabicyclo[8.1.1]dodecane
SMILES [B]1C2[B]C1[B][B][B][B][B][B][B][B]2

Spécifications

Quantity 10 g
Solubility Information Soluble in 10mg/ml chloroform. Insoluble in water.
Formula Weight 144.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material m-Carborane
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only