Learn More
Biotium LysoView™ 540, 1000X in DMSO, Trial Size
Numéro de catalogue. 501964767
Encompass
LysoView™ 540 is a red fluorescent dye that is highly selective for labeling and tracking of lysosomes in live cells, with no wash step required. With absorption and emission at 540/634 nm, the dye can be detected using a Cy3 filter set.
Numéro de catalogue. 501964767 Fournisseur Biotium Code fournisseur 70061T
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
LysoView™ 540 is a red fluorescent dye that is highly selective for labeling and tracking of lysosomes in live cells, with no wash step required. With absorption and emission at 540/634 nm, the dye can be detected using a Cy3 filter set.