Learn More
Biotium LysoView™ 488, 1000X in DMSO, Trial Size
Numéro de catalogue. 501964772
Encompass
LysoView™ 405 is a blue fluorescent lysosome stain for live cells. It has excitation at 318 and 400 nm and emission at 464 nm, for detection in the DAPI or Pacific Blue channels.
Numéro de catalogue. 501964772 Fournisseur Biotium Code fournisseur 70067T
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
LysoView™ 405 is a blue fluorescent lysosome stain for live cells. It has excitation at 318 and 400 nm and emission at 464 nm, for detection in the DAPI or Pacific Blue channels.