Lysyl Endopeptidase cleaves peptide bonds at the carboxy-terminal of both lysine residues and S-aminoethylcysteine residues, making it a valuable tool for analysis of peptide and protein primary structure and for enzymatic synthesis of Lys-X compounds. Also, it can retain complete activity after incubation in 4M urea or in 0.1% SDS solution. Pkg. Size:1 vial (10 AU), Preparation: A 10AU-vial contains Tris-HCl corresponding to 1mL of 2mm Tris HCl buffer (pH 8.0),MW: 27,000 (gel filtration); 30,000 (SDS electrophoresis)