missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lynestrenol 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 163°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 284.44 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Lynestrenol |
Sécurité et manipulation
| RTECSNumber | RC8964300 |
| TSCA | No |