missing translation for 'onlineSavingsMsg'
Learn More

Lycopodium Powder, Reagents

Catalog No. 01337194
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337194 100 g
01337193 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337194 Supplier Labchem Inc Supplier No. C2236000100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lycopodium Powder, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 8023-70-9
Synonym Club moss spores

Specifications

Color Yellowish
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 0.26
Grade Reagent
Physical Form Solid
Chemical Name or Material Lycopodium Powder