missing translation for 'onlineSavingsMsg'
Learn More

Lutetium Standard for ICP/MS - Claritas PPT Grade, SPEX CertiPrep™

Catalog No. CLLU21BY
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLLU21BY 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLLU21BY Supplier SPEX CertiPrep Group LLC Supplier No. CLLU21BY

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7439-94-3
Molecular Formula Lu
Molecular Weight (g/mol) 174.97
MDL Number MFCD00011098
InChI Key OHSVLFRHMCKCQY-UHFFFAOYSA-N
PubChem CID 23929
ChEBI CHEBI:33382
IUPAC Name lutetium
SMILES [Lu]

Specifications

Solvent or Matrix 2% HNO3
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1mg/L
Chemical Name or Material Lutetium Standard for ICP/MS - Claritas PPT Grade
Recommended Storage Ambient