missing translation for 'onlineSavingsMsg'
Learn More

Lutetium ICP Standard, 1000 ppm Lu in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PLU1KN100

Catalog No. PLU1KN-100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Lutetium ICP Standard
12032-20-1 , 7697-37-2 , 7732-18-5
1,000ppm Lu
1 mL = 1 mg Lu
Natural LDPE Bottle
94.0,4.36,0.11
97.0,4.54,0.11

Chemical Identifiers

12032-20-1,7697-37-2,7732-18-5
94.0,4.36,0.11
97.0,4.54,0.11
Lutetium ICP Standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions