CAS RN 12032-20-1
Lutetium(III) oxide, 99.99%, (trace metal basis), -325 mesh
CAS: 12032-20-1 Molecular Formula: Lu2O3 Molecular Weight (g/mol): 397.93 MDL Number: MFCD00011100 InChI Key: UGBIHFMRUDAMBY-UHFFFAOYSA-N Synonym: lutetium 3+ ; oxygen 2-,lutetium oxide,lutetium iii oxide trace rare earth metals basis 1g,cid159406, einecs 234-764-3, lu2o3, lutetium 3+ IUPAC Name: dilutetium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Lu+3].[Lu+3]
Lutetium(III) oxide, REacton™, 99.9% (REO)
CAS: 12032-20-1 Molecular Formula: Lu2O3 Molecular Weight (g/mol): 397.93 MDL Number: MFCD00011100 InChI Key: UGBIHFMRUDAMBY-UHFFFAOYSA-N Synonym: lutetium 3+ ; oxygen 2-,lutetium oxide,lutetium iii oxide trace rare earth metals basis 1g,cid159406, einecs 234-764-3, lu2o3, lutetium 3+ IUPAC Name: dilutetium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Lu+3].[Lu+3]
Lutetium(III) oxide, 99.9% (REO), Thermo Scientific™
CAS: 12032-20-1 Molecular Formula: Lu2O3 Molecular Weight (g/mol): 397.93 MDL Number: MFCD00011100 InChI Key: UGBIHFMRUDAMBY-UHFFFAOYSA-N Synonym: lutetium 3+ ; oxygen 2-,lutetium oxide,lutetium iii oxide trace rare earth metals basis 1g,cid159406, einecs 234-764-3, lu2o3, lutetium 3+ IUPAC Name: dilutetium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Lu+3].[Lu+3]
Lutetium(III) oxide, REacton™, 99.99% (REO)
CAS: 12032-20-1 Molecular Formula: Lu2O3 Molecular Weight (g/mol): 397.93 MDL Number: MFCD00011100 InChI Key: UGBIHFMRUDAMBY-UHFFFAOYSA-N Synonym: lutetium 3+ ; oxygen 2-,lutetium oxide,lutetium iii oxide trace rare earth metals basis 1g,cid159406, einecs 234-764-3, lu2o3, lutetium 3+ IUPAC Name: dilutetium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Lu+3].[Lu+3]
Lutetium AA Standard, 1000 ppm Lu in 3% HNO3, Ricca Chemical
CAS: 12032-20-1
Lutetium ICP Standard, 1000 ppm Lu in 3% HNO3, Ricca Chemical
Chemical Name or Material: Lutetium ICP Standard CAS: 12032-20-1 CAS Min %: 94.0 CAS Max %: 97.0 CAS: 7697-37-2 CAS Min %: 4.36 CAS Max %: 4.54 CAS: 7732-18-5 CAS Min %: 0.11 CAS Max %: 0.11
Lutetium ICP Standard, 10,000 ppm Lu in 5% HNO3, Ricca Chemical
Chemical Name or Material: Lutetium ICP Standard CAS: 12032-20-1 CAS Min %: 89.0 CAS Max %: 92.0 CAS: 7697-37-2 CAS Min %: 8.14 CAS Max %: 8.48 CAS: 7732-18-5 CAS Min %: 1.01 CAS Max %: 1.05