missing translation for 'onlineSavingsMsg'
Learn More

Lutetium ICP Standard, 10,000 ppm Lu in 5% HNO3, Ricca Chemical

Catalog No. PLU10KN100
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PLU10KN100 Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PLU10KN100 Supplier Ricca Chemical Company Supplier No. PLU10KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12032-20-1 , 7697-37-2 , 7732-18-5
CAS Min % 89.0, 8.14, 1.01
CAS Max % 92.0, 8.48, 1.05
Chemical Name or Material Lutetium ICP Standard

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm Lu
Equivalence 1 mL = 10 mg Lu