missing translation for 'onlineSavingsMsg'
Learn More

Lutetium AA Standard, 1000 ppm Lu in 3% HNO3, Ricca Chemical

Catalog No. p-3340517
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
ALU1KN100 Natural LDPE Bottle
ALU1KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ALU1KN100 Supplier Ricca Chemical Company Supplier No. ALU1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12032-20-1 , 7697-37-2 , 7732-18-5

Specifications

Packaging Natural LDPE Bottle
CAS Min % 93.0, 4.56, 0.11
CAS Max % 97.0, 4.74, 0.11
Odor Odorless
Concentration or Composition (by Analyte or Components) 1mL = 1mg Lu (1, 000ppm Lu)Lu2O3 in 3% HNO3
Chemical Name or Material Lutetium AA Standard
ShelfLife 24 months