missing translation for 'onlineSavingsMsg'
Learn More

Lutetium AA Standard, 1000 ppm Lu in 3% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company ALU1KN500

Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. ALU1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

12032-20-1, 7697-37-2, 7732-18-5

Specifications

Lutetium AA Standard
12032-20-1 , 7697-37-2 , 7732-18-5
97.0, 4.74, 0.11
1mL = 1mg Lu (1, 000ppm Lu)Lu2O3 in 3% HNO3
Natural Poly Bottle
93.0, 4.56, 0.11
Odorless
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents

Documents