missing translation for 'onlineSavingsMsg'
Learn More

Luminol 98.0+%, TCI America™

Catalog No. A53011G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53011G 1 g
A530125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A53011G Supplier TCI America Supplier No. A53011G

Chemical Identifiers

CAS 521-31-3
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD00006890
InChI Key HWYHZTIRURJOHG-UHFFFAOYSA-N
Synonym luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide
PubChem CID 10638
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione
SMILES C1=CC2=C(C(=C1)N)C(=O)NNC2=O

Specifications

Melting Point 320°C
Quantity 1 g
Formula Weight 177.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Luminol [Chemiluminescence Reagent]
RTECSNumber TH8890060
TSCA Yes